Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Arlacel(r) A, ACROS Organics™

Click to view available options
Quantity:
25g
100g
Packaging:
Glass bottle
25339-93-9
C24H42O5
MFCD03840565
Mannide monooleate
25339-93-9
C24H42O5
MFCD03840565
Mannide monooleate
Water 0.2% max.
Saponification Value 164 to 172mg KOH/g
Color Colorless
Packaging Glass bottle
Acidity 1mg KOH/g max.
Quantity 25g
Infrared Spectrum Authentic
Formula Weight 410.59
Physical Form Liquid
Chemical Name or Material Mannide monooleate

EINECSNumber : 246-872-8