Learn More
Arlington Scientific Mobile Transport Cart for ASI Lounges
Accessory for Arlington Scientific Lounges
Manufacturer:  Arlington Scientific 20000002
Catalog No. 22-415-168
Add to cart
Description
Description
Portable cart is designed to carry up to five of any of the Arlington Scientific mobile lounges.
  • Can be used as a four-wheel cart or a two-wheel doll
  • Frame is expandable and adjusts to the load size
  • Folds down and retracts for easy storage
Specifications
Specifications
ASI lounges
500 lb.
227kg