Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Arlington Scientific Space Saver Stationary Lounges

Ergonomic design reduces back strain and staff fatigue

Manufacturer:  Arlington ScientificSupplier Diversity Partner LS611004

 View more versions of this product

Catalog No. 22-415-143


  Check Availability
Add to cart

Specifications Description & Specifications

Specifications

650 lb., 294.8kg
Apheresis, oncology, blood collection needs
68.6cm
72 in.
24, 110, 220V
60.96cm
650 lb.
Fabric
72 x 24 x 27 in. (182.8 x 60.96 x 68.6cm)
27 in.
One armrest, clear protective covers and one storage compartment (stores up to 6 cu. ft. of supplies),one center mounted power switch, two aisle facing armrest mounts
182.8cm
24 in.
Yes
294.8kg
Mobile coach
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit