Learn More
Abnova™ ARMCX3 293T Cell Transient Overexpression Lysate (Denatured) (T01)
Human overexpression lysate
Manufacturer:  Abnova™ H00051566T01
Catalog No. 89-935-652
Add to cart
Description
Description
Specifications
Specifications
293T
NM_016607
pCMV-ARMCX3 full-length
100 μL
Human
ARMCX3 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 42.5kDa
Transient overexpression cell lysate was tested with anti-ARMCX3 antibody (Mfr. No. H00051566-B01) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Western Blot