Learn More
Abnova™ ARMCX5 293T Cell Transient Overexpression Lysate (Denatured) (T02)
Human overexpression lysate
Manufacturer:  Abnova™ H00064860T02
Catalog No. 89-935-659
Add to cart
Description
Description
Specifications
Specifications
293T
NM_022838
pCMV-ARMCX5 full-length
100 μL
Human
ARMCX5 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 62.3kDa
Transient overexpression cell lysate was tested with anti-ARMCX5 antibody (Mfr. No. H00064860-B01P) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Western Blot