Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ARMCX6 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP191581
Description
ARMCX6 Polyclonal specifically detects ARMCX6 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
ARMCX6 | |
Polyclonal | |
Unconjugated | |
PBS, 2% Sucrose with 0.09% Sodium Azide | |
armadillo repeat containing, X-linked 6, FLJ20811, protein ARMCX6 | |
Rabbit | |
Protein A purified | |
RUO | |
54470 | |
Centrifuge vial prior to reconstitution. Add 100μL distilled water to a final antibody concentration of 1mg/mL. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
1 mg/ml | |
Western Blot 1.0 ug/ml, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 4-8 ug/ml | |
Q7L4S7 | |
ARMCX6 | |
Synthetic peptide directed towards the N terminal of human ARMCX6 (NP_061880). Peptide sequence TMARPWTEDGDWTEPGAPGGTEDRPSGGGKANRAHPIKQRPFPYEHKNTW. | |
100 μL | |
Primary | |
Expected identity based on immunogen sequence: Mouse: 100%; Human: 100%; Rat: 92%; Canine: 78%. | |
Human, Mouse, Rat, Canine, Equine, Guinea Pig | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction