Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
B7-1/CD80 Antibody (16-10A1), PE/Cy5, Novus Biologicals™
Armenian Hamster Monoclonal Antibody
$409.50 - $603.50
Specifications
Antigen | B7-1/CD80 |
---|---|
Clone | 16-10A1 |
Dilution | Flow Cytometry 1:10-1:1000 |
Applications | Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP14308605
|
Novus Biologicals
NBP1430860.05MG |
0.05 mg |
Each for $409.50
|
|
|||||
NBP14308601
|
Novus Biologicals
NBP1430860.1MG |
0.1 mg |
Each for $603.50
|
|
|||||
Description
B7-1/CD80 Monoclonal specifically detects B7-1/CD80 in Human, Mouse samples. It is validated for Flow Cytometry.Specifications
B7-1/CD80 | |
Flow Cytometry 1:10-1:1000 | |
Monoclonal | |
Armenian Hamster | |
Adaptive Immunity, Cytokine Research, Immunology, Signal Transduction | |
Aqueous buffer, may contain carrier protein/stabilizer (proprietary) with 0.09% Sodium Azide | |
941 | |
The immunogen for this antibody was CD80. | |
Primary | |
Store at 4C. Do not freeze. |
16-10A1 | |
Flow Cytometry | |
PE-Cyanine5 | |
RUO | |
Mouse | |
Activation B7-1 antigen, B7, BB1, CD28LG1, CD28LGB7-1 antigen), CD80 antigen, CD80 molecule, costimulatory factor CD80, costimulatory molecule variant IgV-CD80, CTLA-4 counter-receptor B7.1, T-lymphocyte activation antigen CD80 | |
CD80 | |
IgG | |
Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title