Learn More
Armenian Hamster anti-CCL2/MCP1, Clone: 4E2, Novus Biologicals™
Manufacturer:  Novus Biologicals NBP1432430.5MG
 View more versions of this product
Catalog No. NBP1432435
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
CCL2/MCP1
400
Unconjugated
Aqueous, may contain carrier protein/stabilizer. with 0.09% Sodium Azide
C-C motif chemokine 2, chemokine (C-C motif) ligand 2, GDCF-2, HC11Monocyte secretory protein JE, MCAFMonocyte chemotactic and activating factor, MCP1 Monocyte chemotactic protein 1, MCP-1 Small-inducible cytokine A2, MGC9434, Monocyte chemoattractant protein 1, monocyte chemoattractant protein-1, SCYA2HSMCR30, small inducible cytokine A2 (monocyte chemotactic protein 1, homologous tomouse Sig-je), small inducible cytokine subfamily A (Cys-Cys), member 2, SMC-CF
Armenian Hamster
IgG
0.5 mg
Apoptosis, Cancer, Chemokine Research, Cytokine Research, Hypoxia, Immunology, Innate Immunity, Signal Transduction
Primary
6347
ELISA
0.5 mg/mL
ELISA 1:100-1:2000
Affinity Purified
CCL2
The immunogen for this antibody was CCL2.
Protein A or G purified
RUO
Store at 4C. Do not freeze.
Monoclonal
Mouse