Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fc epsilon RI Antibody (43160), FITC, Novus Biologicals™
Armenian Hamster Monoclonal Antibody
$374.00 - $724.00
Specifications
Antigen | Fc epsilon RI |
---|---|
Clone | 43160 |
Concentration | 0.5 mg/mL |
Dilution | Flow Cytometry < = 0.125 ug/10^6 cells in 100 ul |
Applications | Flow Cytometry |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP14394105
|
Novus Biologicals
NBP1439410.05MG |
0.05 mg |
Each for $374.00
|
|
|||||
NBP14394101
|
Novus Biologicals
NBP1439410.1MG |
0.1 mg |
Each for $603.50
|
|
|||||
NBP1439415
|
Novus Biologicals
NBP1439410.5MG |
0.5 mg |
Each for $724.00
|
|
|||||
Description
Fc epsilon RI Monoclonal specifically detects Fc epsilon RI in Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence.Specifications
Fc epsilon RI | |
0.5 mg/mL | |
Flow Cytometry | |
FITC | |
RUO | |
Mouse | |
alpha polypeptide, Fc fragment of IgE, high affinity I, receptor for; alpha polypeptide, Fc-epsilon RI-alpha, FcERI, high affinity immunoglobulin epsilon receptor alpha-subunit, high affinity immunoglobulin epsilon receptor subunit alpha, high-affinity, of mast cells, alpha polypeptide | |
FCER1A | |
IgG | |
Affinity Purified |
43160 | |
Flow Cytometry < = 0.125 ug/10^6 cells in 100 ul | |
Monoclonal | |
Armenian Hamster | |
Apoptosis, Asthma, Cancer, Immunology, Signal Transduction | |
Aqueous buffer, may contain carrier protein/stabilizer (proprietary) with 0.09% Sodium Azide | |
2205 | |
The immunogen for this antibody was FceRI alpha. | |
Primary | |
Store at 4C. Do not freeze. |
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title