Learn More
Armenian Hamster anti-ICOS, Clone: C398.4A, PE-Cy5, Novus Biologicals™
Manufacturer:  Novus Biologicals NBP1430650.1MG
 View more versions of this product
Catalog No. NBP14306501
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
ICOS
C398.4A
PE-Cyanine5
Affinity Purified
Mouse T cell clone D10.G4.1
Affinity Purified
RUO
Primary
29851
Human, Murine, Rat
Flow Cytometry
0.2mg/mL
Phosphate buffer pH 7.2. Immunogen affinity purified.
Armenian Hamster
IgG
0.1mg
Store at 4°C in the dark.
Monoclonal
CD278 (C398.4A). Reacts with Human, Mouse, Rat. May react with Rhesus and Swine.