Learn More
Armenian Hamster anti-Integrin beta 1/CD29, Clone: HMb1-1, PE-Cy7, Novus Biologicals™
Manufacturer:  Novus Biologicals NBP1429600.1MG
 View more versions of this product
Catalog No. NBP14296001
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Specifications
Specifications
Integrin beta 1/CD29
HMb1-1
Flow Cytometry 1:10-1:1000
Affinity Purified
ITGB1
The immunogen for this antibody was CD29.
Protein A or G purified
RUO
Store at 4C. Do not freeze.
Monoclonal
Mouse
Flow Cytometry
PE-Cyanine7
Aqueous buffer, may contain carrier protein/stabilizer (proprietary) with 0.09% Sodium Azide
CD29, CD29 antigen, Fibronectin receptor subunit beta, FNRBVLAB, GPIIA, integrin beta-1, integrin VLA-4 beta subunit, integrin, beta 1 (fibronectin receptor, beta polypeptide, antigen CD29 includesMDF2, MSK12), MDF2, MSK12, very late activation protein, beta polypeptide, VLA-4 subunit beta, VLA-BETA
Armenian Hamster
IgG
0.1 mg
Cancer, Cytokine Research, Cytoskeleton Markers, Embryonic Stem Cell Markers, Immunology, Mesenchymal Stem Cell Markers, Neuronal Cell Markers, Neuronal Stem Cell Markers, Signal Transduction, Stem Cell Markers
Primary
3688