Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Armenian Hamster anti-LAIR1, DyLight 488, Clone: 113, Novus Biologicals™

Armenian Hamster Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP143324G

Catalog No. NBP143324G


Add to cart

Description

Description

Ensure accurate, reproducible results in Western Blot, Flow Cytometry, Immunoprecipitation

LAIR1 Monoclonal antibody specifically detects LAIR1 in Mouse samples. It is validated for Western Blot, Flow Cytometry.
Specifications

Specifications

LAIR1
113
DyLight 488
50mM Sodium Borate with 0.05% Sodium Azide
CD305 antigen, CD305leukocyte-associated Ig-like receptor 1, hLAIR1, LAIR-1, leukocyte-associated immunoglobulin-like receptor 1
Armenian Hamster
IgG
0.1 ml
Store at 4C in the dark.
Monoclonal
Mouse
Flow Cytometry, Western Blot
1mg/mL
Western Blot, Flow Cytometry
Affinity Purified
LAIR1
The immunogen for this antibody was CD305.
Protein A or G purified
RUO
Primary
3903
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only