Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Armenian Hamster IgG, Clone: 299Arm, PE-Cy5, Isotype control, Novus Biologicals™

Armenian Hamster Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP1430710.05MG

 View more versions of this product

Catalog No. NBP14307105


Add to cart

Description

Description

IgG Monoclonal antibody specifically detects IgG in Armenian Hamster samples. It is validated for Flow Cytometry.
Specifications

Specifications

IgG
299Arm
Flow Cytometry 1:10 - 1:1000
Conjugated
Armenian Hamster
0.05 mg
Store at 4C in the dark.
Monoclonal
Armenian Hamster
Flow Cytometry
PE-Cyanine5
Aqueous buffer, may contain carrier protein/stabilizer (proprietary). with 0.09% Sodium Azide
immunoglobulin heavy constant gamma 1 (G1m marker)
IgG
RUO
Primary
This 299Arm antibody has been tested by flow cytometric analysis of mouse splenocytes and normal human peripheral blood. This should be used at the same concentration as the experimental antibody.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit