Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ARMET Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA520432
Description
A suggested positive control is rat brain tissue lysate. PA5-20432 can be used with blocking peptide PEP-0549.
The protein encoded by this gene is localized in the endoplasmic reticulum and golgi, and is also secreted. Reducing expression of this gene increases susceptibility to ER stress-induced death and promotes cell proliferation. The protein was initially thought to be longer at the N-terminus and to contain an arginine-rich region but transcribed evidence indicates a smaller open reading frame that does not encode the arginine tract. The presence of polymorphisms in the arginine-rich region, including a specific mutation that changes the previously numbered codon 50 from ATG to AGG, have been reported in a variety of solid tumors; however, these polymorphisms were later shown to exist in normal tissues and are thus not tumor-related.
Specifications
ARMET | |
Polyclonal | |
Unconjugated | |
MANF | |
3230402M22Rik; AA407711; AA408789; AA673178; Arginine-rich protein; arginine-rich, mutated in early stage tumors; ARMET; ARP; D17914; D18Mgi17; Manf; mesencephalic astrocyte derived neurotrophic factor; mesencephalic astrocyte-derived neurotrophic factor; MGC142148; MGC142150; Protein ARMET | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
315989, 74840, 7873 | |
Maintain refrigerated at 2°C to 8°C for up to 3 months. For long term storage store at -20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunohistochemistry (PFA fixed), Western Blot | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
P0C5H9, P55145, Q9CXI5 | |
MANF | |
a 12 amino acid peptide from near the amino terminus of human MANF. | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction