Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ARMET Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA527609
Description
Recommended positive controls: 293T, HepG2, Molt-4, Raji. Predicted reactivity: Mouse (99%), Zebrafish (84%), Pig (100%), Bovine (98%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The protein encoded by this gene is localized in the endoplasmic reticulum and golgi, and is also secreted. Reducing expression of this gene increases susceptibility to ER stress-induced death and promotes cell proliferation. The protein was initially thought to be longer at the N-terminus and to contain an arginine-rich region but transcribed evidence indicates a smaller open reading frame that does not encode the arginine tract. The presence of polymorphisms in the arginine-rich region, including a specific mutation that changes the previously numbered codon 50 from ATG to AGG, have been reported in a variety of solid tumors; however, these polymorphisms were later shown to exist in normal tissues and are thus not tumor-related.
Specifications
ARMET | |
Polyclonal | |
Unconjugated | |
MANF | |
3230402M22Rik; AA407711; AA408789; AA673178; Arginine-rich protein; arginine-rich, mutated in early stage tumors; ARMET; ARP; D17914; D18Mgi17; Manf; mesencephalic astrocyte derived neurotrophic factor; mesencephalic astrocyte-derived neurotrophic factor; MGC142148; MGC142150; Protein ARMET | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
7873 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 40% glycerol and 0.01% thimerosal; pH 7 | |
P55145 | |
MANF | |
Recombinant fragment corresponding to a region within amino acids 25 and 182 of Human ARMET. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction