Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ARMT1 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP310729100UL
Description
ARMT1 Polyclonal specifically detects ARMT1 in Human samples. It is validated for Western Blot.Specifications
ARMT1 | |
Polyclonal | |
Western Blot 1.0 ug/ml | |
Rabbit | |
Affinity purified | |
RUO | |
79624 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
PBS buffer, 2% sucrose | |
The immunogen is a synthetic peptide directed towards the N-terminal region of Human C6orf211 (NP_078849). Peptide sequence SDGKSRWFYSPWLLVECYMYRRIHEAIIQSPPIDYFDVFKESKEQNFYGS | |
100 μg | |
Primary | |
Human | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction