Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ARMX2 Polyclonal Antibody
ARMX2 Polyclonal Antibody
Supplier: Thermo Scientific PA598529
Description
ARMX2 Polyclonal Antibody for Western Blot, IHC (P), ELISA

Specifications
ARMX2 | |
Polyclonal | |
Unconjugated | |
ARMCX2 | |
3230401N03Rik; AI043003; ALEX2; ALEX2 protein; ARM protein lost in epithelial cancers on chromosome X 2; armadillo repeat containing, X-linked 2; armadillo repeat protein ALEX2; armadillo repeat-containing X-linked protein 2; ARMCX2; GASP9; Kiaa0512; Protein ALEX2 | |
Rabbit | |
Protein G | |
RUO | |
9823 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
5 mg/mL | |
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
Q7L311 | |
ARMCX2 | |
Recombinant Human Armadillo repeat-containing X-linked protein 2 protein (161-460AA). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction