Learn More
Tocris Bioscience™ ARN 272
FAAH-like anandamide transporter (FLAT) inhibitor
Manufacturer:  Tocris Bioscience™ 4574/10
Catalog No. 45-741-0
Add to cart
This item is not returnable. View return policy
Description
Description
FAAH-like anandamide transporter (FLAT) inhibitor (IC50 = 1.8 μM); cytosolic variant of FAAH-1 that binds anandamide. Attenuates anandamide internalization and deactivation in vitro and in vivo respectively. Exhibits analgesic effects in rodent models of CB1-mediated nociceptive and inflammatory pain.
Specifications
Specifications
ARN 272
>98
10mg
ARN 272
432.47