Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ARNTL2 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$106.19 - $320.71

Specifications

Antigen ARNTL2
Dilution Western Blot 1.0 ug/ml, Immunohistochemistry
Applications Western Blot, Immunohistochemistry
Conjugate Unconjugated
Format Purified
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB125915
View Documents
Novus Biologicals
NBP310507100UL
100 μg
Each for $320.71
Add to cart
 
NB125916
View Documents
Novus Biologicals
NBP31050725UL
25 μg
Each for $106.19
Add to cart
 
Description

Description

ARNTL2 Polyclonal antibody specifically detects ARNTL2 in Human samples. It is validated for Western Blot, Immunohistochemistry
Specifications

Specifications

ARNTL2
Western Blot, Immunohistochemistry
Purified
RUO
PBS buffer, 2% sucrose
56938
Polyclonal
Affinity purified
Western Blot 1.0 ug/ml, Immunohistochemistry
Unconjugated
Rabbit
Human
Aryl hydrocarbon receptor nuclear translocator-like protein 2, Basic-helix-loop-helix-PAS protein MOP9, bHLHe6, BMAL2, Brain and muscle ARNT-like 2, Class E basic helix-loop-helix protein 6, CLIF, CYCLE-like factor, Member of PAS protein 9, MOP9, PAS domain-containing protein 9, PASD9, Transcription Factor BMAL2
The immunogen is a synthetic peptide directed towards the N terminal region of human ARNTL2 (NP_064568). Peptide sequence PTAMGSFSSHMTEFPRKRKGSDSDPSQSGIMTEKVVEKLSQNPLTYLLST
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit