Learn More
Aromatase Antibody (H4), FITC, Novus Biologicals™
SDP
Mouse Monoclonal Antibody
Supplier:  Novus Biologicals NBP261939F
Catalog No. NBP261939F
Add to Cart
Description
Description
Aromatase Monoclonal antibody specifically detects Aromatase in Human, Rat, Porcine samples. It is validated for Immunohistochemistry, Immunocytochemistry, Immunofluorescence.
Specifications
Specifications
Aromatase
Monoclonal
FITC
PBS with 0.05% Sodium Azide
CYP19A1
Synthetic peptide corresponding to AAs 376-390 of human aromatase (KALEDDVIGYPVKK)
0.1 mL
Primary
Human, Rat, Porcine
Purified
Immunohistochemistry, Immunocytochemistry, Immunofluorescence
H4
Immunohistochemistry, Immunocytochemistry/Immunofluorescence
ARO1EC 1.14.14.1, Aromatase, AROsubfamily XIX (aromatization of androgens), CYARMGC104309, CYPXIX, cytochrome P450, family 19, subfamily A, polypeptide 1, Cytochrome P-450AROM, Estrogen synthase, microsomal monooxygenase
Mouse
Protein A purified
Breast Cancer, Cancer, Cell Biology, Cell Cycle and Replication, Chromatin Research, Innate Immunity, Lipid and Metabolism, Neuroscience
1588
Store at 4C in the dark.
IgG2a
Recommendations
Recommendations