Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aromatase Mouse anti-Human, Clone: CYP19A1/4257, Novus Biologicals™

Mouse Monoclonal Antibody

$138.37 - $352.89

Specifications

Antigen Aromatase
Clone CYP19A1/4257
Concentration 0.2 mg/ml
Dilution Western Blot 1-2 ug/ml, ELISA, Immunohistochemistry-Paraffin 1-2 ug/ml
Applications Western Blot, ELISA, Immunohistochemistry (Paraffin)
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB123125
View Documents
Novus Biologicals
NBP30782610UG
100 μg
Each for $352.89
Add to cart
 
NB123126
View Documents
Novus Biologicals
NBP30782620UG
20 μg
Each for $138.37
Add to cart
 
Description

Description

Aromatase Monoclonal antibody specifically detects Aromatase in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry (Paraffin)
Specifications

Specifications

Aromatase
0.2 mg/ml
Western Blot, ELISA, Immunohistochemistry (Paraffin)
Purified
RUO
Human
ARO1EC 1.14.14.1, Aromatase, AROsubfamily XIX (aromatization of androgens), CYARMGC104309, CYPXIX, cytochrome P450, family 19, subfamily A, polypeptide 1, Cytochrome P-450AROM, Estrogen synthase, microsomal monooxygenase
Synthetic peptide corresponding to aa376-390 of human Aromatase (KALEDDVIGYPVKK)
Monoclonal
Protein A or G purified
CYP19A1/4257
Western Blot 1-2 ug/ml, ELISA, Immunohistochemistry-Paraffin 1-2 ug/ml
Unconjugated
Mouse
Breast Cancer, Cancer, Cell Biology, Cell Cycle and Replication, Chromatin Research, Innate Immunity, Lipid and Metabolism, Neuroscience
10 mM PBS with 0.05% BSA
1588
IgG2a
Primary
Store at 4°C.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit