Learn More
Aromatase Mouse anti-Human, Porcine, DyLight 350, Clone: H4, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP261939UV
Catalog No. NBP261939UV
Add to cart
Description
Description
Aromatase Monoclonal antibody specifically detects Aromatase in Human, Porcine samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
Aromatase
H4
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
50mM Sodium Borate with 0.05% Sodium Azide
1588
Mouse
IgG2a
Primary
0.1 ml
Store at 4C in the dark.
Western Blot,Immunohistochemistry,Immunocytochemistry,Immunofluorescence
DyLight 350
Purified
ARO1EC 1.14.14.1, Aromatase, AROsubfamily XIX (aromatization of androgens), CYARMGC104309, CYPXIX, cytochrome P450, family 19, subfamily A, polypeptide 1, Cytochrome P-450AROM, Estrogen synthase, microsomal monooxygenase
CYP19A1
Synthetic peptide corresponding to AAs 376-390 of human aromatase (KALEDDVIGYPVKK)
Monoclonal
Protein A purified
Breast Cancer, Cancer, Cell Biology, Cell Cycle and Replication, Chromatin Research, Innate Immunity, Lipid and Metabolism, Neuroscience
Human,Porcine