Learn More
Medchemexpress LLC Arotinolol | 68377-92-4 | 99.9% | 25 MG

Supplier: Medchemexpress LLC HY122537A25MG
Arotinolol is an α/β-adrenergic receptor blocker provided as a solid research chemical for laboratory use. The compound (CAS 68377-92-4) is supplied with high purity and defined storage recommendations.
- High purity suitable for research applications (99.9%).
- Molecular weight 371.54 g/mol and formula C15H21N3O2S3.
- Appearance: off-white to light yellow solid.
- Recommended storage: powder -20 °C (3 years) or 4 °C (2 years); in solvent -80 °C (6 months) or -20 °C (1 month).
- Available in small vial sizes for research flexibility.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.