Learn More
ARPC2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 150581AP
Catalog No. 50-172-9940
Add to cart
Description
Description
Tumor necrosis factor (TNF) receptor associated factors (TRAFs) were initially discovered as adaptor proteins that link the TNF receptor superfamily to signaling pathways and are thus important regulators of cell death and cellular response to stress. TRAF proteins share a homology region that allows them to bind to cell receptors and other TRAF proteins, causing the activation of different signal cascades depending on the TRAFs involved. For example, TRAF2 and TRAF3 directly bind to the CD40, a NF receptor superfamily member involved in inducing B cell immunity, and are critical for NF-kappa-B activation in mouse B lymphocytes. TRAF2 along with TRAF6 has also been shown to be required for CD40 signaling in nonhemopoietic cells. TRAF2 also interacts with the TRFR superfamily member lymphotoxin-beta receptor (LTbetaR) in association with TRAF3 and the apoptosis inhibitors cIAP1 and Smac.
Specifications
Specifications
ARPC2
0.23 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O15144, P85970, Q9CVB6
ARC34, Arp2/3 complex 34 kDa subunit, ARPC2, p34 Arc, PNAS 139, PRO2446
Rabbit
IgG
150 μL
-20°C
Primary
10109, 301511, 76709
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
ARPC2
Liquid
Arpc2
ARPC2 Fusion Protein Ag7041
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat