Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ARPC5 Mouse anti-Human, Clone: OTI2G1, liquid, TrueMAB
Mouse Monoclonal Antibody
Manufacturer: OriGene TA505696
Description
This gene encodes one of seven subunits of the human Arp2/3 protein complex. The Arp2/3 protein complex has been implicated in the control of actin polymerization in cells and has been conserved through evolution. The exact role of the protein encoded by this gene, the p16 subunit, has yet to be determined.Specifications
ARPC5 | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
O15511 | |
ARC16, dJ127C7.3, p16-Arc | |
Mouse | |
IgG1 | |
100 μL | |
-20° C, Avoid Freeze/Thaw Cycles | |
Primary | |
10092 |
Immunohistochemistry (Paraffin), Western Blot | |
OTI2G1 | |
Unconjugated | |
ARPC5 | |
Liquid | |
ARPC5 | |
Full length human recombit protein of human ARPC5 produced in HEK293T cell. | |
Affinity Chromatography | |
RUO | |
Antibody | |
Monoclonal | |
Human |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title