Learn More
ARPC5L Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 220251AP
Catalog No. 50-173-3691
Add to cart
Description
Description
May function as component of the Arp2/3 complex which is involved in regulation of actin polymerization and together with an activating nucleation-promoting factor (NPF) mediates the formation of branched actin networks.
Specifications
Specifications
ARPC5L
0.23 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
A1L108, Q9BPX5, Q9D898
ARC16 2, ARPC5L
Rabbit
IgG
150 μL
-20°C
Primary
296710, 74192, 81873
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation
Unconjugated
ARPC5L
Liquid
Arpc5l
ARPC5L Fusion Protein Ag16881
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat