Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Arpin Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00
Specifications
Antigen | Arpin |
---|---|
Dilution | Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500 |
Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
Arpin Polyclonal antibody specifically detects Arpin in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
Arpin | |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Human | |
Q7Z6K5 | |
348110 | |
IgG | |
Immunogen affinity purified |
Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
Polyclonal | |
Purified | |
RUO | |
PBS (pH 7.2), 40% Glycerol | |
2610034B18Rik, C15orf38, RIKEN cDNA 2610034B18 gene | |
This antibody was developed against Recombinant Protein corresponding to amino acids: EVMELELGAGVRLKTRGDGPFLDSLAKLEAGTVTKCNFTGDGKTGASWTDNIMAQKCSKGAAAEIREQGDGAEDEEWDD | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title