Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ARRB2 Mouse anti-Human, Rat, Unconjugated, Clone: 4D2, Abnova™
Mouse Monoclonal Antibody.
Supplier: Abnova Corporation H00000409M06J
Description
ARRB2 Monoclonal antibody specifically detects ARRB2 in Human, Rat samples. It is validated for WB-Ce, IF, S-ELISA, ELISA, WB-Re.Specifications
ARRB2 | |
Monoclonal | |
Unconjugated | |
ARRB2 | |
ARB2, ARR2, BARR2, DKFZp686L0365 | |
ARRB2 (AAH07427.1, 300 a.a. ∼ 409 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa | |
Primary | |
Human, Rat | |
IgG2a κ |
Immunofluorescence, ELISA, Western Blot | |
4D2 | |
arrestin, beta 2 | |
BC007427.2 | |
Mouse | |
100 μg | |
409 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction