Learn More
ARRDC3 Rabbit anti-Human, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 250601AP
Catalog No. 50-173-4702
Add to cart
Specifications
Specifications
ARRDC3
0.18 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q96B67
ARRDC3, arrestin domain containing 3, KIAA1376, TLIMP
Rabbit
IgG
150 μL
-20°C
Primary
57561
Immunocytochemistry, Immunofluorescence, Western Blot
Unconjugated
ARRDC3
Liquid
ARRDC3
ARRDC3 Fusion Protein Ag18717
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human