Learn More
Sklar InstrumentsSupplier Diversity Partner Forceps, Arruga Capsule Forceps, Sklar 4 in., Angled, Smooth, Stainless Steel, EA. DFS Item
Manufacturer:  Sklar InstrumentsSupplier Diversity Partner 665340
Forceps, Arruga Capsule Forceps, Sklar, 4 in., Angled, Smooth, Stainless Steel, EA. Sklar's Arruga Capsule Forceps are an ophthalmic instrument used for holding the lens capsule during cataract procedures. The tips are specially designed so they do not damage the iris. They are oval shaped with a slight cup. This product has a standard curve and overall length of 4 inches. Lifetime warranty against all manufacturer defects.
Catalog No. 50-111-8955