Alfa Aesar™ Arsenazo III disodium salt

Manufacturer: Alfa Aesar™ A1183806

Quantity 5g

 View more versions of this product

Catalog No. AAA1183806

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 62337-00-2
    Molecular Formula C22H16As2N4Na2O14S2
    Formula Weight 820.34
    MDL Number MFCD00003936
    UN Number UN3465
    Synonym 3,6-Bis(2,2'-arsonophenylazo)-4,5-dihydroxy-2,7-naphthalenedisulfonic acid disodium salt
    Chemical Name or Material Arsenazo III disodium salt

    Specifications

    Formula Weight 820.34
    Health Hazard 3 GHS P Statement
    P261-P301+P310-P304+P340-P311-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Call a POISON CENTER or doctor/physician.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    MDL Number MFCD00003936
    CAS 62337-00-2
    UN Number UN3465
    DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ORGANOARSENIC COMPOUND, SOLID, N.O.S.
    Synonym 3,6-Bis(2,2'-arsonophenylazo)-4,5-dihydroxy-2,7-naphthalenedisulfonic acid disodium salt
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H301-H331
    Toxic if swallowed.
    Toxic if inhaled.
    Quantity 5g
    Molecular Formula C22H16As2N4Na2O14S2
    Melting Point >320°C
    Chemical Name or Material Arsenazo III disodium salt

    Description

    Applications
    Arsenazo III disodium salt is a diazo dye reagent used in the spectrophotometric determination of aluminum, gallium and indium. Further, it is used in the complexometric determination of lanthanides, thallium, scandium thorium, uranium and yttrium. In addition to this, it acts as a metal indicator and form specifically colored metal ion complexes in aqueous media.

    Solubility
    Soluble in water.

    Notes
    Incompatible with strong oxidizing agents.

    GHS H Statement
    H301-H331
    Toxic if swallowed.
    Toxic if inhaled.

    GHS P Statement
    P261-P301+P310-P304+P340-P311-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Call a POISON CENTER or doctor/physician.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit