Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Arsenazo III Disodium Salt, Spectrum™

Click to view available options
Quantity:
5 mg
25 mg
1 g
5 g
Packaging:
Amber Glass Bottle
62337-00-2
62337-00-2
Packaging Amber Glass Bottle
Quantity 1 g
CAS Max % 1
Grade Reagent

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.