Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Arsenic(III) chloride, 99.999% (metals basis)

Click to view available options
Quantity:
25g
100g
7784-34-1
AsCl3
181.272
MFCD00014165
OEYOHULQRFXULB-UHFFFAOYSA-N
trichloroarsine,arsenic trichloride,arsenic iii chloride,arsenic chloride,arsenous trichloride,arsenic butter,arsenous chloride,arsenious chloride,butter of arsenic,chlorure arsenieux
24570
CHEBI:63952
trichloroarsane
Cl[As](Cl)Cl
7784-34-1
AsCl3
181.272
MFCD00014165
OEYOHULQRFXULB-UHFFFAOYSA-N
trichloroarsine,arsenic trichloride,arsenic iii chloride,arsenic chloride,arsenous trichloride,arsenic butter,arsenous chloride,arsenious chloride,butter of arsenic,chlorure arsenieux
24570
CHEBI:63952
trichloroarsane
Cl[As](Cl)Cl
UN Number UN1560
Merck Index 14,801
Formula Weight 181.27
Physical Form Liquid
Percent Purity 99.999%
Density 2.163 g/mL
Odor Acrid
Sensitivity Moisture sensitive
Boiling Point 130.2°C
Color Brown-Yellow to Colorless
Melting Point -16°C
Quantity 100g
Chemical Name or Material Arsenic(III) chloride
Assay (metals basis)
Show More Show Less

GHS H Statement
H300-H331-H350-H314
Fatal if swallowed.
Toxic if inhaled.
May cause cancer.
Causes severe skin burns and eye damage.

P264b-P270-P301+P310-P330-P501c

H300

DOTInformation : Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: ARSENIC TRICHLORIDE

EINECSNumber : 232-059-5

RTECSNumber : CG1750000

TSCA : Yes

Recommended Storage : Ambient temperatures