Alfa Aesar™ Arsenic(III) chloride, Reagent Grade

Manufacturer: Alfa Aesar™ 3149918

Quantity 50g

 View more versions of this product

Catalog No. AA3149918

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 7784-34-1
    Formula Weight 181.27
    MDL Number MFCD00014165
    UN Number UN1560
    Merck Index 14,801
    Synonym Arsenic trichloride
    Chemical Name or Material Arsenic(III) chloride
    Grade Reagent Grade

    Specifications

    Formula Weight 181.27
    Health Hazard 3 GHS P Statement
    P273-P201-P309-P310-P501a
    Avoid release to the environment.
    Obtain special instructions before use.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Solubility Information Decomposes in water with formation of As(OH)3 and HCl; Increasing dilutions in water result in As2O3 precipitation
    RTECS Number CG1750000
    Sensitivity Moisture Sensitive
    Merck Index 14,801
    Synonym Arsenic trichloride
    Boiling Point 130.2°C
    MDL Number MFCD00014165
    CAS 7784-34-1
    UN Number UN1560
    Density 2.163g/mL
    DOT Information Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: ARSENIC TRICHLORIDE
    TSCA TSCA
    Odor Acrid
    Melting Point −16°C
    Quantity 50g
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H300-H331-H350-H314
    Fatal if swallowed.
    Toxic if inhaled.
    May cause cancer.
    Causes severe skin burns and eye damage.
    Physical Form Liquid
    Chemical Name or Material Arsenic(III) chloride
    Grade Reagent Grade

    GHS H Statement
    H300-H331-H350-H314
    Fatal if swallowed.
    Toxic if inhaled.
    May cause cancer.
    Causes severe skin burns and eye damage.

    GHS P Statement
    P273-P201-P309-P310-P501a
    Avoid release to the environment.
    Obtain special instructions before use.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit