Alfa Aesar™ Arsenic(III) ethoxide

Manufacturer: Alfa Aesar™ 3149809

Quantity 10g

 View more versions of this product

Catalog No. AA3149809

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 3141-12-6
    Formula Weight 210.07
    MDL Number MFCD00015120
    UN Number UN2929
    Synonym Triethoxyarsine
    Chemical Name or Material Arsenic(III) ethoxide

    Specifications

    Formula Weight 210.07
    Health Hazard 3 GHS P Statement
    P210-P261-P301+P310-P303+P361+P353-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    MDL Number MFCD00015120
    Health Hazard 1 Danger
    Boiling Point 166°C
    Sensitivity Moisture Sensitive
    DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC LIQUIDS, FLAMMABLE, ORGANIC, N.O.S.
    RTECS Number CG5210000
    Physical Form Liquid
    CAS 3141-12-6
    UN Number UN2929
    Synonym Triethoxyarsine
    Density 1.21g/mL
    Quantity 10g
    Health Hazard 2 GHS H Statement
    H301-H331-H226
    Toxic if swallowed.
    Toxic if inhaled.
    Flammable liquid and vapor.
    Chemical Name or Material Arsenic(III) ethoxide

    GHS H Statement
    H301-H331-H226
    Toxic if swallowed.
    Toxic if inhaled.
    Flammable liquid and vapor.

    GHS P Statement
    P210-P261-P301+P310-P303+P361+P353-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit