Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Arsenic(III) ethoxide

Manufacturer:  Alfa Aesar™ 3149818

Quantity 50g

 View more versions of this product

Catalog No. AA3149818

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications Description & Specifications

Chemical Identifiers

3141-12-6
MFCD00015120
Triethoxyarsine
210.07
UN2929
Arsenic(III) ethoxide

Specifications

3141-12-6
210.07
UN2929
1.21g/mL
50g
Arsenic(III) ethoxide
Liquid
MFCD00015120
Triethoxyarsine
166°C
Moisture Sensitive

Safety and Handling

SDS & Safety Documents

GHS H Statement
H301-H331-H226
Toxic if swallowed.
Toxic if inhaled.
Flammable liquid and vapor.

GHS P Statement
P210-P261-P301+P310-P303+P361+P353-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC LIQUIDS, FLAMMABLE, ORGANIC, N.O.S.

RTECSNumber : CG5210000

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit