Alfa Aesar™ Arsenic(III) iodide, 98%

Manufacturer: Alfa Aesar™ 4506718

Quantity 50g

 View more versions of this product

Catalog No. AA4506718

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7784-45-4
    Molecular Formula AsI3
    Formula Weight 455.64
    MDL Number MFCD00014170
    UN Number UN1557
    Merck Index 14,803
    Synonym Arsenic triiodide
    Chemical Name or Material Arsenic (III) iodide

    Specifications

    Formula Weight 455.64
    Health Hazard 3 GHS P Statement
    P261-P301+P310-P304+P340-P311-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Call a POISON CENTER or doctor/physician.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    TSCA TSCA
    Odor Pungent
    Sensitivity Moisture Sensitive
    CAS 7784-45-4
    UN Number UN1557
    Health Hazard 1 Danger
    Density 4.39g/mL
    DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ARSENIC COMPOUNDS, SOLID, N.O.S.
    Physical Form Small Pieces
    Molecular Formula AsI3
    MDL Number MFCD00014170
    Merck Index 14,803
    Synonym Arsenic triiodide
    Boiling Point 424°C
    Melting Point 140°C
    Quantity 50g
    Health Hazard 2 GHS H Statement
    H301-H331
    Toxic if swallowed.
    Toxic if inhaled.
    Chemical Name or Material Arsenic (III) iodide
    Assay 98%

    GHS H Statement
    H301-H331
    Toxic if swallowed.
    Toxic if inhaled.

    GHS P Statement
    P261-P301+P310-P304+P340-P311-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Call a POISON CENTER or doctor/physician.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit