Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Arsenic(III) oxide, 99.5%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
500g
Packaging:
Glass bottle
1327-53-3
O3As2
MFCD00003433
Arsenic trioxide
1327-53-3
O3As2
MFCD00003433
Arsenic trioxide
Assay Percent Range 99.5%
Linear Formula As2O3
Antimony (Sb) 500ppm max.
Formula Weight 197.84
Physical Form Powder
Percent Purity ≥99.4%
Chloride 50ppm max.
Insoluble Matter 0.2% max. (in dilute HCl)
Iron (Fe) 20ppm max.
Lead (Pb) 20ppm max.
Packaging Glass bottle
Color White to Beige
Quantity 100g
Chemical Name or Material Arsenic(III) oxide
Show More Show Less

GHS P Statement Causes severe skin burns and eye damage. Very toxic to aquatic life with long lasting effects. May cause cancer. Fatal if swallowed.

GHS P Statement IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Avoid release to the environment. Obtain special instructions before use. IF exposed or concerned: Get medical advice/attention.

Danger