Learn More
Arsenic(III) oxide, primary standard, ACS, 99.95-100.05%, Thermo Scientific Chemicals
Description
Used in the manufacturing of such products as glass and enamels. Arsenic trioxide is utilized in various studies on cellular processes, notably in investigations of leukemia and principal material for the production of many arsenic compounds.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Boiling Point | 465°C (sublimation) |
Melting Point | 312.3°C |
Physical Form | Powder |
Quantity | 25 g |
Assay Percent Range | 99.95-100.05% |
UN Number | UN1561 |
Merck Index | 14,804 |
Solubility Information | Soluble in dilute acids and alkalies. Insoluble in organic solvents. Slightly soluble in water. |
Formula Weight | 197.84 |
Percent Purity | 99.95 to 100.05% |
Show More |
Safety and Handling
Hazard Category | H300-H314-H335-H350-H371 |
Hazard Statement | GHS H Statement H300-H350-H314-H318 |
Precautionary Statement | P201-P202-P260-P264b-P270-P271-P281-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c |
DOTInformation | Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ARSENIC TRIOXIDE |
EINECSNumber | 215-481-4 |
RTECSNumber | CG3325000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.