Alfa Aesar™ Arsenic(III) oxide, Puratronic™, 99.996% (metals basis excluding Sb), Sb 20ppm max

Manufacturer: Alfa Aesar™ 1117309

Quantity 10g

 View more versions of this product

Catalog No. AA1117309

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1327-53-3
    Formula Weight 197.84
    MDL Number MFCD00003433
    UN Number UN1561
    Merck Index 14,804
    Chemical Name or Material Arsenic(III) oxide
    Grade Puratronic™

    Specifications

    Formula Weight 197.84
    Health Hazard 3 GHS P Statement
    P260-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Physical Form Powder
    Health Hazard 2 GHS H Statement
    H300-H350-H314-H318
    Fatal if swallowed.
    May cause cancer.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    TSCA TSCA
    Odor Odorless
    CAS 1327-53-3
    Quantity 10g
    Health Hazard 1 Danger
    UN Number UN1561
    Boiling Point 465°C (subl.)
    Density 3.74g/mL
    DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ARSENIC TRIOXIDE
    MDL Number MFCD00003433
    Merck Index 14,804
    Melting Point 312.3°C
    RTECS Number CG3325000
    Chemical Name or Material Arsenic(III) oxide
    Assay 99.996%, (metals basis excluding Sb), Sb 20ppm max.
    Grade Puratronic™

    GHS H Statement
    H300-H350-H314-H318
    Fatal if swallowed.
    May cause cancer.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P260-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit