Alfa Aesar™ Arsenic(III) selenide, 99.999% (metals basis)

For vacuum deposition

Manufacturer: Alfa Aesar™ 1284906

Quantity 5g

 View more versions of this product

Catalog No. AA1284906

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1303-36-2
    Formula Weight 386.72
    MDL Number MFCD00014166
    UN Number UN3077
    Merck Index 14,805
    Synonym Arsenic triselenide
    Chemical Name or Material Arsenic(III) selenide

    Specifications

    Formula Weight 386.72
    Health Hazard 3 GHS P Statement
    P260-P284-P304+P340-P320-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Wear respiratory protection.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Specific treatment is urgent (see label).
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Health Hazard 2 GHS H Statement
    H330
    Fatal if inhaled.
    RTECS Number CG2285000
    TSCA TSCA
    Odor Odorless
    Physical Form Fused Pieces
    UN Number UN3077
    Melting Point 260°C
    Quantity 5g
    Health Hazard 1 Danger
    Particle Size 1 to 6mm
    MDL Number MFCD00014166
    Merck Index 14,805
    Synonym Arsenic triselenide
    Density 4.75g/mL
    DOT Information Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
    CAS 1303-36-2
    Chemical Name or Material Arsenic(III) selenide
    Assay 99.999%, (metals basis)

    GHS H Statement
    H330
    Fatal if inhaled.

    GHS P Statement
    P260-P284-P304+P340-P320-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Wear respiratory protection.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Specific treatment is urgent (see label).
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit