Alfa Aesar™ Arsenic(III) telluride, 99.999% (metals basis)

Manufacturer: Alfa Aesar™ 4170814

Quantity 25g

 View more versions of this product

Catalog No. AA4170814

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 12044-54-1
    Formula Weight 532.64
    MDL Number MFCD00014167
    UN Number UN1557
    Chemical Name or Material Arsenic(III) telluride

    Specifications

    Formula Weight 532.64
    Health Hazard 3 GHS P Statement
    P261-P301+P310-P304+P340-P311-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Call a POISON CENTER or doctor/physician.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Mesh Size −160 Mesh
    MDL Number MFCD00014167
    CAS 12044-54-1
    Quantity 25g
    Health Hazard 2 GHS H Statement
    H301-H331
    Toxic if swallowed.
    Toxic if inhaled.
    Physical Form Powder
    Melting Point 381°C
    Health Hazard 1 Danger
    Density 6.50g/mL
    DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ARSENIC COMPOUNDS, SOLID, N.O.S.
    TSCA TSCA
    Odor Odorless
    UN Number UN1557
    Chemical Name or Material Arsenic(III) telluride
    Assay 99.999%, (metals basis)

    GHS H Statement
    H301-H331
    Toxic if swallowed.
    Toxic if inhaled.

    GHS P Statement
    P261-P301+P310-P304+P340-P311-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Call a POISON CENTER or doctor/physician.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit