Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Arsenic, Lump, Purified, Spectrum™

Click to view available options
Quantity:
25 g
125 g
500 g
2.5 kg
Packaging:
Amber Glass Bottle
7440-38-2
7440-38-2
Grade Purified
CAS Max % 1
Packaging Amber Glass Bottle
Quantity 2.5 kg

Caution: For manufacturing, processing or repacking. Read and understand the label and Safety Data Sheet (SDS) prior to use.