Arsenic Polycrystalline lump, 2-8mm (0.08-0.3 in.), Puratronic™, 99.9999% (Metals basis), Alfa Aesar™

Manufacturer: Alfa Aesar 1337522

Quantity 100g

 View more versions of this product

Catalog No. AA1337522

    N/A

Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-766-7000 or send an email to CS.WebEmail@thermofisher.com for assistance. 


    Chemical Identifiers

    CAS 7440-38-2
    Physical Form Polycrystalline Lump, 2-8mm (0.08-0.3 in.)
    Assay Percent Range 99.9999% (Metals basis)
    MDL Number MFCD00085309
    Chemical Name or Material Arsenic

    Specifications

    Physical Form Polycrystalline Lump, 2-8mm (0.08-0.3 in.)
    Quantity 100g
    Color Gray
    DOT Information DOT Class: 6.1, Packaging Group: II
    EINECS Number 213-148-6
    Odor Odorless
    Assay Percent Range 99.9999% (Metals basis)
    Appearance Gray lump
    MDL Number MFCD00085309
    CAS 7440-38-2
    Health Hazard 1 H301-H331-H400-H410; Toxic if swallowed. Toxic if inhaled. Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects.
    Chemical Name or Material Arsenic

    H301-H331-H400-H410; Toxic if swallowed. Toxic if inhaled. Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit