Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Arsenic sponge, 30mm (1.18 in.) & down, 99.5% (metals basis)

Click to view available options
Quantity:
25 g
100 g
7440-38-2
As
74.922
MFCD00085309
RQNWIZPPADIBDY-UHFFFAOYSA-N
arsenicals,arsenic, inorganic,arsenic-75,arsen,gray,grey,black,metallic,compounds,colloidal
5359596
CHEBI:27563
arsenic
[As]
7440-38-2
As
74.922
MFCD00085309
RQNWIZPPADIBDY-UHFFFAOYSA-N
arsenicals,arsenic, inorganic,arsenic-75,arsen,gray,grey,black,metallic,compounds,colloidal
5359596
CHEBI:27563
arsenic
[As]
Quantity 100 g
UN Number UN1558
Odor Odorless
Physical Form Irregular Pieces
Particle Size 30mm (1.18 in.) & down
Percent Purity 99.5%
Chemical Name or Material Arsenic sponge
Assay (metals basis)

GHS H Statement
H301-H331
Toxic if swallowed.
Toxic if inhaled.

P201-P202-P261-P264b-P270-P271-P281-P301+P310-P304+P340-P308+P313-P311-P330-P501c

H301+H331-H350

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ARSENIC

EINECSNumber : 213-148-6

TSCA : Yes

Recommended Storage : Ambient temperatures