Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Arsenic Trioxide, Powder, Technical, Spectrum™

Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
45 kg
50 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
1327-53-3
1327-53-3
Grade Technical
CAS Max % 1
Packaging Fiber Drum
Quantity 45 kg

CAUTION: For manufacturing or laboratory use only. Not for pesticide use. Read and understand the label and Safety Data Sheet (SDS) prior to use.