Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Arsenic (V) Standard, Stock Solution, 1.00 mL = 1.00 mg As (V), APHA, Spectrum™

Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Amber Glass Bottle
1303-28-2,7732-18-5,1310-73-2
1303-28-2,7732-18-5,1310-73-2
Packaging Amber Glass Bottle
Quantity 100 mL

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.