Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Arsenious Acid Solution, For Iodide, APHA, Spectrum™

Click to view available options
Quantity:
500 mL
Packaging:
Poly Bottle
CAS 1327-53-3
Grade Chemical Solution
Packaging Poly Bottle
Quantity 500 mL