Learn More
ARSI Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA549984
Catalog No. PA549984
Add to cart
Description
Description
The antibody detects endogenous levels of total ARSI protein.
Sulfatases (EC 3.1.5.6), such as ARSI, hydrolyze sulfate esters from sulfated steroids, carbohydrates, proteoglycans, and glycolipids. They are involved in hormone biosynthesis, modulation of cell signaling, and degradation of macromolecules
Specifications
Specifications
ARSI
Polyclonal
Unconjugated
ARSI
Liquid
ARSI
Synthesized peptide derived from internal of human ARSI.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Immunocytochemistry, Immunofluorescence, Western Blot
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Q5FYB1
arylsulfatase I, ASI, EC 3.1.6.-,
Rabbit
IgG
100 μL
-20°C
Primary
340075