Learn More
Abnova™ ARSK 293T Cell Transient Overexpression Lysate (Denatured)
Human overexpression lysate
Manufacturer:  Abnova™ H00153642T01
Catalog No. 89-935-757
Add to cart
Description
Description
Specifications
Specifications
293T
NM_198150.2
pCMV-ARSK full-length
100 μL
Human
ARSK 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 59.07kDa
Transient overexpression cell lysate was tested with anti-ARSK antibody (Mfr. No. H00153642-B01) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Western Blot